Assay Details
| Gene Symbol: DIMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285600.2 | NP_001272529.1 | 7 - 8 | 628 | 59 | |
| GenBank mRNA | JN942971.1 | - | 7 - 8 | 628 | 59 | |
| JT749293.1 | - | 7 - 8 | 707 | 59 | ||
More Information
Set Membership: |
|

DIMT1
Genomic Map