Assay Details
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285619.1 | NP_001272548.1 | 5 - 6 | 882 | 71 | |
GenBank mRNA | AF325499.1 | - | 5 - 6 | 1078 | 71 | |
AF339909.1 | - | 5 - 6 | 1054 | 71 | ||
GU947654.1 | - | 5 - 6 | 882 | 71 | ||
JO634430.2 | - | 2 - 3 | 3500 | 71 | ||
JX854036.1 | - | 5 - 6 | 882 | 71 | ||
KA308300.1 | - | 5 - 6 | 1317 | 71 | ||
KT315581.1 | - | 5 - 6 | 983 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
