Assay Details
| Gene Symbol: SPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285667.1 | NP_001272596.1 | 5 - 6 | 519 | 64 | |
| XM_013964504.2 | XP_013819958.1 | 6 - 7 | 640 | 64 | ||
| GenBank mRNA | EU295699.1 | - | 5 - 6 | 519 | 64 | |
| KF699859.1 | - | 6 - 7 | 569 | 64 | ||
| KJ789112.1 | - | 5 - 6 | 560 | 64 | ||
More Information
Set Membership: |
|

SPP1
Genomic Map