Assay Details
| Gene Symbol: PSMA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285698.1 | NP_001272627.1 | 5 - 6 | 625 | 117 | |
| GenBank mRNA | FJ594753.1 | - | 5 - 6 | 625 | 117 | |
| JO580480.1 | - | 2 - 3 | 386 | 117 | ||
More Information
Set Membership: |
|

PSMA6
Genomic Map