Assay Details
| Gene Symbol: CMPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314253.1 | NP_001301182.1 | 2 - 3 | 333 | 68 | |
| GenBank mRNA | JQ818260.1 | - | 2 - 3 | 333 | 68 | |
| JT731577.1 | - | 5 - 6 | 1370 | 68 | ||
More Information
Set Membership: |
|

CMPK1
Genomic Map