Assay Details
Gene Symbol: FABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287558.1 | NP_001274487.1 | 1 - 2 | 73 | 65 | |
GenBank mRNA | HQ606485.1 | - | 1 - 2 | 83 | 65 | |
JO420454.2 | - | 3 - 4 | 390 | 65 | ||
JQ031286.1 | - | 1 - 2 | 73 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
