Assay Details
| Gene Symbol: LPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285607.1 | NP_001272536.1 | 5 - 6 | 780 | 60 | |
| XM_013966067.2 | XP_013821521.2 | 5 - 6 | 1083 | 60 | ||
| GenBank mRNA | DQ370053.1 | - | 5 - 6 | 903 | 60 | |
| DQ997818.1 | - | 5 - 6 | 780 | 60 | ||
| GU082383.1 | - | 5 - 6 | 780 | 60 | ||
| JQ670882.1 | - | 5 - 6 | 922 | 60 | ||
More Information
Set Membership: |
|

LPL
Genomic Map