Assay Details
Gene Symbol: SLC39A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285772.1 | NP_001272701.1 | 1 - 2 | 274 | 66 | |
XM_005685384.3 | XP_005685441.1 | 1 - 2 | 702 | 66 | ||
GenBank mRNA | HQ615881.1 | - | 1 - 2 | 274 | 66 | |
KA336899.1 | - | 3 - 4 | 1093 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
