Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285772.1 | NP_001272701.1 | 2 - 3 | 404 | 92 | |
| GenBank mRNA | HQ615881.1 | - | 2 - 3 | 404 | 92 | |
| KA336899.1 | - | 2 - 3 | 963 | 92 | ||
More Information
Set Membership: |
|

SLC39A2
Genomic Map