Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285772.1 | NP_001272701.1 | 3 - 4 | 461 | 78 | |
| GenBank mRNA | HQ615881.1 | - | 3 - 4 | 461 | 78 | |
| KA336899.1 | - | 1 - 2 | 906 | 78 | ||
More Information
Set Membership: |
|

SLC39A2
Genomic Map