Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287569.1 | NP_001274498.1 | 4 - 5 | 327 | 64 | |
GenBank mRNA | JO431072.2 | - | 4 - 5 | 1576 | 64 | |
JQ407044.1 | - | 4 - 5 | 327 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
