Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314259.1 | NP_001301188.1 | 2 - 3 | 409 | 66 | |
| XM_005681305.3 | XP_005681362.1 | 3 - 4 | 850 | 66 | ||
| XM_018049158.1 | XP_017904647.1 | 4 - 5 | 688 | 66 | ||
| GenBank mRNA | KP721342.1 | - | 2 - 3 | 409 | 66 | |
More Information
Set Membership: |
|

PITX2
Genomic Map