Assay Details
| Gene Symbol: FOXM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314325.1 | NP_001301254.1 | 5 - 6 | 1101 | 61 | |
| GenBank mRNA | KP056826.1 | - | 5 - 6 | 1101 | 61 | |
More Information
Set Membership: |
|

FOXM1
Genomic Map