Assay Details
Gene Symbol: S-LZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285711.1 | NP_001272640.1 | 1 - 2 | 133 | 90 | |
GenBank mRNA | GQ903681.1 | - | 1 - 2 | 133 | 90 | |
JO340487.2 | - | 2337 | 90 | |||
KC954666.1 | - | 1 - 2 | 133 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
