Assay Details
| Gene Symbol: EEF1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314314.1 | NP_001301243.1 | 4 - 5 | 407 | 113 | |
| GenBank mRNA | JP351417.2 | - | 4 - 5 | 535 | 113 | |
| JQ308785.1 | - | 4 - 5 | 407 | 113 | ||
More Information
Set Membership: |
|

EEF1B2
Genomic Map