Assay Details
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060005.1 | XP_017915494.1 | 7 - 8 | 1908 | 59 | |
GenBank mRNA | JO435613.2 | - | 2 - 3 | 436 | 59 | |
JO435714.2 | - | 2 - 3 | 436 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
