Assay Details
| Gene Symbol: HOXC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679865.3 | XP_005679922.1 | 1 - 2 | 514 | 55 | |
| XM_018047654.1 | XP_017903143.1 | 2 - 3 | 917 | 55 | ||
More Information
Set Membership: |
|

HOXC6
Genomic Map