Assay Details
| Gene Symbol: LOC102177846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059034.1 | XP_017914523.1 | 3 - 4 | 450 | 92 | |
| GenBank mRNA | JO353219.2 | - | 3 - 4 | 479 | 92 | |
More Information
Set Membership: |
|

LOC102177846
Genomic Map