Assay Details
Gene Symbol: ADORA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962542.2 | XP_013817996.1 | 1 - 2 | 598 | 71 | |
GenBank mRNA | JO576174.1 | - | 1 - 2 | 841 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
