Assay Details
| Gene Symbol: NCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050814.1 | XP_017906303.1 | 3 - 4 | 1071 | 81 | |
| XM_018050821.1 | XP_017906310.1 | 3 - 4 | 1074 | 81 | ||
| XM_018050830.1 | XP_017906319.1 | 3 - 4 | 1045 | 81 | ||
| XM_018050839.1 | XP_017906328.1 | 3 - 4 | 1054 | 81 | ||
| XM_018050844.1 | XP_017906333.1 | 2 - 3 | 830 | 81 | ||
| GenBank mRNA | JO610951.2 | - | 1 - 2 | 620 | 81 | |
More Information
Set Membership: |
|

NCK1
Genomic Map