Assay Details
Gene Symbol: LOC102184534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968612.2 | XP_013824066.2 | 2 - 3 | 457 | 71 | |
GenBank mRNA | JO538955.2 | - | 2 - 3 | 503 | 71 | |
JO632805.2 | - | 2 - 3 | 482 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
