Assay Details
| Gene Symbol: PGRMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044217.1 | XP_017899706.1 | 2 - 3 | 789 | 106 | |
| GenBank mRNA | JO420698.2 | - | 2 - 3 | 588 | 106 | |
| JO634286.2 | - | 2 - 3 | 488 | 106 | ||
More Information
Set Membership: |
|

PGRMC1
Genomic Map