Assay Details
Gene Symbol: LOC102190507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682628.2 | XP_005682685.2 | 2 - 3 | 192 | 84 | |
XM_013965276.1 | XP_013820730.1 | 2 - 3 | 204 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
