Assay Details
| Gene Symbol: FBXL21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682960.3 | XP_005683017.2 | 2 - 3 | 674 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXL21P| Gene Symbol: FBXL21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682960.3 | XP_005683017.2 | 2 - 3 | 674 | 114 | |
Set Membership: |
|