Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680008.3 | XP_005680065.1 | 4 - 5 | 698 | 62 | |
| GenBank mRNA | JO363342.2 | - | 1 - 2 | 1344 | 62 | |
| KA338569.1 | - | 4 - 5 | 720 | 62 | ||
More Information
Set Membership: |
|

ARF3
Genomic Map