Assay Details
| Gene Symbol: RTP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675140.3 | XP_005675197.2 | 1 - 2 | 282 | 60 | |
| GenBank mRNA | JO403984.2 | - | 1 - 2 | 197 | 60 | |
More Information
Set Membership: |
|

RTP4
Genomic Map