Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699919.3 | XP_005699976.1 | 3 - 4 | 534 | 57 | |
| XM_013976022.2 | XP_013831476.1 | 3 - 4 | 371 | 57 | ||
| XM_018043208.1 | XP_017898697.1 | 3 - 4 | 402 | 57 | ||
| GenBank mRNA | JO612075.2 | - | 7 - 8 | 1052 | 57 | |
More Information
Set Membership: |
|

BRMS1
Genomic Map