Assay Details
| Gene Symbol: SERPINH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059336.1 | XP_017914825.1 | 2 - 3 | 802 | 73 | |
| XM_018059337.1 | XP_017914826.1 | 3 - 4 | 817 | 73 | ||
| GenBank mRNA | JO367257.2 | - | 3 - 4 | 1327 | 73 | |
| JO409892.2 | - | 2 - 3 | 769 | 73 | ||
More Information
Set Membership: |
|

SERPINH1
Genomic Map