Assay Details
| Gene Symbol: NECTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064224.1 | XP_017919713.1 | 4 - 5 | 1071 | 59 | |
| XM_018064281.1 | XP_017919770.1 | 4 - 5 | 1071 | 59 | ||
| GenBank mRNA | JO361414.2 | - | 2 - 3 | 382 | 59 | |
| JO397501.2 | - | 2 - 3 | 932 | 59 | ||
| JO444202.2 | - | 4 - 5 | 810 | 59 | ||
| JO444979.2 | - | 4 - 5 | 810 | 59 | ||
| KA340304.1 | - | 2 - 3 | 623 | 59 | ||
More Information
Set Membership: |
|

NECTIN3
Genomic Map