Assay Details
| Gene Symbol: LOC102189116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682625.3 | XP_005682682.1 | 14 - 15 | 1723 | 77 | |
| XM_013965331.2 | XP_013820785.1 | 12 - 13 | 1449 | 77 | ||
| XM_013965333.2 | XP_013820787.1 | 13 - 14 | 1515 | 77 | ||
| XM_018050726.1 | XP_017906215.1 | 14 - 15 | 1722 | 77 | ||
| XM_018050727.1 | XP_017906216.1 | 14 - 15 | 1792 | 77 | ||
| XM_018050728.1 | XP_017906217.1 | 15 - 16 | 1827 | 77 | ||
| XM_018050729.1 | XP_017906218.1 | 14 - 15 | 1713 | 77 | ||
| XM_018050730.1 | XP_017906219.1 | 14 - 15 | 1716 | 77 | ||
| GenBank mRNA | JO427600.1 | - | 17 - 18 | 2129 | 77 | |
| JO514642.2 | - | 2 - 3 | 570 | 77 | ||
More Information
Set Membership: |
|

LOC102189116
Genomic Map