Assay Details
| Gene Symbol: LOC102189116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682625.3 | XP_005682682.1 | 15 - 16 | 1821 | 68 | |
| XM_013965331.2 | XP_013820785.1 | 13 - 14 | 1547 | 68 | ||
| XM_013965333.2 | XP_013820787.1 | 14 - 15 | 1613 | 68 | ||
| XM_018050726.1 | XP_017906215.1 | 15 - 16 | 1820 | 68 | ||
| XM_018050727.1 | XP_017906216.1 | 15 - 16 | 1890 | 68 | ||
| XM_018050728.1 | XP_017906217.1 | 16 - 17 | 1925 | 68 | ||
| XM_018050729.1 | XP_017906218.1 | 15 - 16 | 1811 | 68 | ||
| XM_018050730.1 | XP_017906219.1 | 15 - 16 | 1814 | 68 | ||
| GenBank mRNA | JO427600.1 | - | 18 - 19 | 2227 | 68 | |
| JO514642.2 | - | 1 - 2 | 472 | 68 | ||
More Information
Set Membership: |
|

LOC102189116
Genomic Map