Assay Details
Gene Symbol: LOC102169480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685235.3 | XP_005685292.3 | 1 - 2 | 322 | 62 |
Gene Symbol: CD82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO581298.1 | - | 2593 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
