Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971210.2 | XP_013826664.1 | 5 - 6 | 506 | 88 | |
XM_013971211.2 | XP_013826665.1 | 5 - 6 | 503 | 88 | ||
XM_013971212.2 | XP_013826666.1 | 3 - 4 | 260 | 88 | ||
GenBank mRNA | KA314555.1 | - | 1 - 2 | 216 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
