Assay Details
Gene Symbol: LSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696141.3 | XP_005696198.1 | 2 - 3 | 218 | 85 | |
XM_018066885.1 | XP_017922374.1 | 2 - 3 | 218 | 85 | ||
GenBank mRNA | JO622828.2 | - | 2 - 3 | 356 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
