Assay Details
Gene Symbol: ELAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039723.1 | XP_017895212.1 | 2 - 3 | 243 | 79 | |
XM_018039724.1 | XP_017895213.1 | 2 - 3 | 519 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
