Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698833.3 | XP_005698890.1 | 15 - 16 | 3306 | 79 | |
XM_018041786.1 | XP_017897275.1 | 15 - 16 | 2895 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
