Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698833.3 | XP_005698890.1 | 3 - 4 | 1291 | 68 | |
XM_018041786.1 | XP_017897275.1 | 3 - 4 | 880 | 68 | ||
XM_018041787.1 | XP_017897276.1 | 3 - 4 | 1075 | 68 | ||
XM_018041788.1 | XP_017897277.1 | 3 - 4 | 880 | 68 | ||
GenBank mRNA | JO578840.1 | - | 7 - 8 | 1455 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
