Assay Details
| Gene Symbol: RBPJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057917.1 | XP_017913406.1 | 2 - 3 | 2749 | 61 | |
| XM_018057919.1 | XP_017913408.1 | 2 - 3 | 2749 | 61 | ||
More Information
Set Membership: |
|

RBPJL
Genomic Map