Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065708.1 | XP_017921197.1 | 2 - 3 | 514 | 73 | |
| XM_018065709.1 | XP_017921198.1 | 2 - 3 | 383 | 73 | ||
| XM_018065710.1 | XP_017921199.1 | 2 - 3 | 198 | 73 | ||
| XM_018065713.1 | XP_017921202.1 | 2 - 3 | 163 | 73 | ||
| XM_018065714.1 | XP_017921203.1 | 2 - 3 | 381 | 73 | ||
| XM_018065715.1 | XP_017921204.1 | 2 - 3 | 219 | 73 | ||
| GenBank mRNA | JO604746.2 | - | 6 - 7 | 1983 | 73 | |
More Information
Set Membership: |
|

ZNF131
Genomic Map