Assay Details
| Gene Symbol: LOC102170015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974709.2 | XP_013830163.2 | 3 - 4 | 531 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102170015| Gene Symbol: LOC102170015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974709.2 | XP_013830163.2 | 3 - 4 | 531 | 80 | |
Set Membership: |
|