Assay Details
Gene Symbol: LOC102189098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680483.3 | XP_005680540.2 | 1 | 1028 | 123 | |
XM_005680485.3 | XP_005680542.1 | 2 | 953 | 123 |
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045034.1 | XP_017900523.1 | 11 - 12 | 1033 | 123 | |
XM_018045035.1 | XP_017900524.1 | 9 - 10 | 961 | 123 | ||
XM_018045037.1 | XP_017900526.1 | 10 - 11 | 958 | 123 | ||
GenBank mRNA | JO352040.2 | - | 1 - 2 | 278 | 123 | |
JO412933.2 | - | 2 - 3 | 1053 | 123 | ||
JO412934.2 | - | 2 - 3 | 1053 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
