Assay Details
| Gene Symbol: COL8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674830.2 | XP_005674887.1 | 3 - 4 | 615 | 65 | |
| XM_018058129.1 | XP_017913618.1 | 4 - 5 | 664 | 65 | ||
| GenBank mRNA | JO382381.2 | - | 1 - 2 | 481 | 65 | |
| JO442740.2 | - | 1 - 2 | 686 | 65 | ||
| JO534439.2 | - | 1 - 2 | 1579 | 65 | ||
| JO627341.2 | - | 1 - 2 | 482 | 65 | ||
| KA331987.1 | - | 3 - 4 | 509 | 65 | ||
More Information
Set Membership: |
|

COL8A1
Genomic Map