Assay Details
| Gene Symbol: ADPRHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041561.1 | XP_017897050.1 | 3 - 4 | 542 | 69 | |
| GenBank mRNA | JO422582.2 | - | 3 - 4 | 466 | 69 | |
More Information
Set Membership: |
|

ADPRHL2
Genomic Map