Assay Details
| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047106.1 | XP_017902595.1 | 3 - 4 | 1720 | 78 | |
| GenBank mRNA | JO437490.2 | - | 3 - 4 | 552 | 78 | |
| JT688190.1 | - | 6 - 7 | 1571 | 78 | ||
More Information
Set Membership: |
|

ZNRF2
Genomic Map