Assay Details
Gene Symbol: SCIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693445.3 | XP_005693502.1 | 2 - 3 | 245 | 85 | |
XM_005693446.3 | XP_005693503.1 | 2 - 3 | 1127 | 85 | ||
XM_013972056.2 | XP_013827510.1 | 2 - 3 | 245 | 85 | ||
GenBank mRNA | JO603801.2 | - | 2 - 3 | 237 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
