Assay Details
| Gene Symbol: HDDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694987.2 | XP_005695044.1 | 3 - 4 | 464 | 70 | |
| GenBank mRNA | JO444054.2 | - | 1 - 2 | 452 | 70 | |
| JO613715.2 | - | 1 - 2 | 449 | 70 | ||
More Information
Set Membership: |
|

HDDC3
Genomic Map