Assay Details
| Gene Symbol: GPANK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696586.3 | XP_005696643.1 | 3 - 4 | 1194 | 76 | |
| XM_005696590.3 | XP_005696647.1 | 2 - 3 | 959 | 76 | ||
| XM_013974025.2 | XP_013829479.1 | 3 - 4 | 1616 | 76 | ||
| XM_018038611.1 | XP_017894100.1 | 3 - 4 | 1406 | 76 | ||
| XM_018038612.1 | XP_017894101.1 | 3 - 4 | 1125 | 76 | ||
| XM_018038613.1 | XP_017894102.1 | 3 - 4 | 1107 | 76 | ||
| GenBank mRNA | KA347289.1 | - | 3 - 4 | 1580 | 76 | |
More Information
Set Membership: |
|

GPANK1
Genomic Map