Assay Details
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040111.1 | XP_017895600.1 | 3 - 4 | 775 | 51 | |
XM_018040112.1 | XP_017895601.1 | 3 - 4 | 775 | 51 | ||
XM_018040113.1 | XP_017895602.1 | 2 - 3 | 388 | 51 | ||
GenBank mRNA | JO600082.2 | - | 1528 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
