Assay Details
| Gene Symbol: LOC108633183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043754.1 | XP_017899243.1 | 1310 | 62 | ||
| XM_018043755.1 | XP_017899244.1 | 1229 | 62 | |||
| Gene Symbol: LOC102173827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045521.1 | XP_017901010.1 | 2 - 3 | 331 | 62 | |
| GenBank mRNA | JO407132.2 | - | 1 - 2 | 88 | 62 | |
More Information
Set Membership: |
|

LOC108633183
Genomic Map