Assay Details
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053661.1 | XP_017909150.1 | 3 - 4 | 534 | 83 | |
XM_018053662.1 | XP_017909151.1 | 3 - 4 | 333 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
